Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYB3R2
Entrez Gene ID
827977
Gene Name
myb domain protein 3r2
Gene Aliases
AT4G00540, ATMYB3R2, C-MYB-LIKE TRANSCRIPTION FACTOR 3R-2, F6N23.19, F6N23_19, MYB DOMAIN PROTEIN 3R2, MYB3R-2, PC-MYB2, myb domain protein 3r2
Chromosome Location
Chr.4: 234597 - 237582 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116278.1 NP_567179.1 7 - 8 1108 112
GenBank mRNA AF151647.1 - 7 - 8 882 112
AF218054.2 - 7 - 8 1108 112

View Full Product Details