Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SK32
Entrez Gene ID
828023
Gene Name
shaggy-like protein kinase 32
Gene Aliases
ASKTHETA, AT4G00720, ATSK32, F6N23.11, F6N23_11, SHAGGY-LIKE PROTEIN KINASE THETA, shaggy-like protein kinase 32
Chromosome Location
Chr.4: 293588 - 297479 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116297.4 NP_191981.1 1 - 2 542 60
GenBank mRNA AK226199.1 - 1 - 2 361 60
AY062099.1 - 1 - 2 354 60
AY124881.1 - 1 - 2 65 60
Y07822.1 - - 263 60

View Full Product Details