Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANL2
Entrez Gene ID
828022
Gene Name
Homeobox-leucine zipper family protein / lipid-binding START domain-containing protein
Gene Aliases
AHDP, ANTHOCYANINLESS 2, ARABIDOPSIS THALIANA HOMEODOMAIN PROTEIN, AT4G00730, F6N23.10, F6N23_10, HOMEODOMAIN PROTEIN AHDP
Chromosome Location
Chr.4: 299261 - 304612 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116298.4 NP_567183.2 7 - 8 2178 83
GenBank mRNA AK226968.1 - 7 - 8 2019 83

View Full Product Details