The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001340272.1 | NP_001328370.1 | 2 - 3 | 433 | 96 |
NM_116311.4 | NP_567194.1 | 2 - 3 | 485 | 96 | |
GenBank mRNA | AF378866.1 | - | 2 - 3 | 223 | 96 |
AY052732.1 | - | 2 - 3 | 155 | 96 | |
AY102641.1 | - | 2 - 3 | 262 | 96 |