Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCB5
- Entrez Gene ID
-
826974
- Gene Name
- P-glycoprotein 5
- Gene Aliases
-
AT4G01830, ATP-binding cassette B5, P-glycoprotein 5, PGP5, T7B11.9, T7B11_9
- Chromosome Location
-
Chr.4: 785551 - 790447 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_116413.2 |
NP_192092.1 |
1 - 2 |
216 |
81 |
View Full Product Details