Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCB5
Entrez Gene ID
826974
Gene Name
P-glycoprotein 5
Gene Aliases
AT4G01830, ATP-binding cassette B5, P-glycoprotein 5, PGP5, T7B11.9, T7B11_9
Chromosome Location
Chr.4: 785551 - 790447 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340339.1 NP_001329375.1 5 - 6 2398 62
NM_116413.2 NP_192092.1 9 - 10 3042 62

View Full Product Details