Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MOS6
Entrez Gene ID
827472
Gene Name
ARM repeat superfamily protein
Gene Aliases
6, AT4G02150, ATIMPALPHA3, IMPA-3, IMPORTIN ALPHA 3, IMPORTIN ALPHA ISOFORM 3, MODIFIER OF SNC1, T10M13.16, T10M13_16
Chromosome Location
Chr.4: 950546 - 953690 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340367.1 NP_001328074.1 1 - 2 305 68
NM_116447.3 NP_192124.1 1 - 2 305 68
GenBank mRNA AF385693.1 - 1 - 2 304 68
AY081725.1 - 1 - 2 217 68
DQ087970.1 - 1 - 2 217 68

View Full Product Details