Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSH1
Entrez Gene ID
828096
Gene Name
RELA/SPOT homolog 1
Gene Aliases
AT-RSH1, AT4G02260, ATRSH1, RELA-SPOT HOMOLOG 1, RELA/SPOT HOMOLOG 1, RELA/SPOT homolog 1, T2H3.10, T2H3_10
Chromosome Location
Chr.4: 985123 - 991506 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340369.1 NP_001329795.1 14 - 15 1638 86
NM_116459.4 NP_567226.1 14 - 15 1655 86
NM_178956.4 NP_849287.2 14 - 15 1649 86
NM_202772.2 NP_974501.1 14 - 15 1646 86
GenBank mRNA AF225702.1 - 14 - 15 1499 86

View Full Product Details