Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUS3
Entrez Gene ID
828081
Gene Name
sucrose synthase 3
Gene Aliases
AT4G02280, ATSUS3, T2H3.8, T2H3_8, sucrose synthase 3
Chromosome Location
Chr.4: 994726 - 998991 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116461.4 NP_192137.1 2 - 3 673 72
GenBank mRNA AY051001.1 - 2 - 3 447 72
AY056784.1 - 2 - 3 472 72
AY142511.1 - 2 - 3 233 72
KJ138769.1 - 2 - 3 233 72

View Full Product Details