Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK22
Entrez Gene ID
825806
Gene Name
calcium-dependent kinase-like protein
Gene Aliases
AT4G04710, T4B21.12, T4B21_12, calcium-dependent protein kinase 22
Chromosome Location
Chr.4: 2390307 - 2392989 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340499.1 NP_001329212.1 5 - 6 1186 110

View Full Product Details