Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLS
Entrez Gene ID
825825
Gene Name
cardiolipin synthase
Gene Aliases
AT4G04870, T4B21.19, T4B21_19, cardiolipin synthase
Chromosome Location
Chr.4: 2461674 - 2464150 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116725.5 NP_567273.1 2 - 3 699 78
GenBank mRNA AY059735.1 - 2 - 3 633 78
AY091269.1 - 2 - 3 553 78

View Full Product Details