Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ISA3
Entrez Gene ID
826481
Gene Name
isoamylase 3
Gene Aliases
AT4G09020, ATISA3, F23J3.50, F23J3_50, isoamylase 3
Chromosome Location
Chr.4: 5783909 - 5789100 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116971.6 NP_192641.2 6 - 7 886 103
GenBank mRNA AK227049.1 - 6 - 7 723 103
AY091058.1 - 6 - 7 719 103
AY133739.1 - 6 - 7 696 103

View Full Product Details