Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNL2
Entrez Gene ID
832081
Gene Name
GNOM-like 2
Gene Aliases
AT5G19610, GNOM-like 2, T29J13.30, T29J13_30
Chromosome Location
Chr.5: 6617665 - 6622104 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121966.2 NP_197462.1 1 - 2 544 136

View Full Product Details