Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GPCR
- Entrez Gene ID
-
841725
- Gene Name
- G protein coupled receptor
- Gene Aliases
-
AT1G52920, F14G24.19, F14G24_19, G protein coupled receptor, G-PROTEIN COUPLED RECEPTOR 2, GCR2
- Chromosome Location
-
Chr.1: 19709275 - 19711081 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104170.3 |
NP_175700.2 |
5 - 6 |
922 |
70 |
View Full Product Details