Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLP7
Entrez Gene ID
841766
Gene Name
tubby like protein 7
Gene Aliases
AT1G53320, AtTLP7, F12M16.22, F12M16_22, tubby like protein 7
Chromosome Location
Chr.1: 19890935 - 19893504 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104210.4 NP_564627.1 1 - 2 395 124
GenBank mRNA AF370146.1 - 1 - 2 382 124
AY059088.1 - 1 - 2 320 124
AY092403.1 - 1 - 2 320 124

View Full Product Details