Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PME2
Entrez Gene ID
841820
Gene Name
pectin methylesterase 2
Gene Aliases
AT1G53830, ATPME2, T18A20.6, T18A20_6, pectin methylesterase 2
Chromosome Location
Chr.1: 20098522 - 20100978 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104260.5 NP_175786.1 1 - 2 648 72
GenBank mRNA AF361637.1 - 1 - 2 634 72
AY133609.1 - 1 - 2 608 72

View Full Product Details