Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bZIP4
Entrez Gene ID
842243
Gene Name
basic leucine-zipper 4
Gene Aliases
AT1G59530, ATBZIP4, T30E16.6, T30E16_6, basic leucine-zipper 4
Chromosome Location
Chr.1: 21868190 - 21868708 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104646.2 NP_176162.1 1 - 1 462 98
GenBank mRNA AB493509.1 - 1 - 1 390 98
AF400619.1 - 1 - 1 390 98
DQ446373.1 - 1 - 1 390 98

View Full Product Details