Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPAC43
Entrez Gene ID
842356
Gene Name
RNA polymerase I subunit 43
Gene Aliases
AT1G60620, ATRPAC43, RNA POLYMERASE I SUBUNIT 43, RNA polymerase I subunit 43
Chromosome Location
Chr.1: 22330894 - 22333568 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333901.1 NP_001320426.1 4 - 5 760 104
NM_104745.3 NP_176261.1 3 - 4 804 104
GenBank mRNA AF386969.1 - 3 - 4 561 104
BT001161.1 - 3 - 4 473 104
L34773.1 - 3 - 4 592 104

View Full Product Details