Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPSF73-I
Entrez Gene ID
842393
Gene Name
cleavage and polyadenylation specificity factor 73-I
Gene Aliases
AT1G61010, T7P1.15, T7P1_15, cleavage and polyadenylation specificity factor 73-I
Chromosome Location
Chr.1: 22474267 - 22477765 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036138.2 NP_001031215.1 4 - 5 542 123
NM_001333924.1 NP_001319278.1 4 - 5 542 123
NM_001333925.1 NP_001321253.1 4 - 5 536 123
NM_104782.4 NP_176297.1 4 - 5 536 123
NM_179504.2 NP_849835.1 4 - 5 542 123
GenBank mRNA AK316692.1 - 4 - 5 479 123
AK316794.1 - 4 - 5 507 123
AY150478.1 - 4 - 5 437 123

View Full Product Details