Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SMO1-1
- Entrez Gene ID
-
826819
- Gene Name
- sterol-4alpha-methyl oxidase 1-1
- Gene Aliases
-
AT4G12110, ATSMO1, ATSMO1-1, F16J13.180, F16J13_180, STEROL 4-ALPHA-METHYL-OXIDASE 1, sterol-4alpha-methyl oxidase 1-1
- Chromosome Location
-
Chr.4: 7254029 - 7256406 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117281.3 |
NP_192948.1 |
4 - 5 |
1011 |
82 |
GenBank mRNA |
AY125499.1 |
- |
4 - 5 |
989 |
82 |
View Full Product Details