Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC1B
Entrez Gene ID
826820
Gene Name
Sec1/munc18-like (SM) proteins superfamily
Gene Aliases
AT4G12120, ATSEC1B, F16J13.190, F16J13_190
Chromosome Location
Chr.4: 7256522 - 7261180 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117282.3 NP_567388.1 4 - 5 560 107

View Full Product Details