Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GH9C2
Entrez Gene ID
842747
Gene Name
glycosyl hydrolase 9C2
Gene Aliases
AT1G64390, AtGH9C2, F15H21.9, F15H21_9, glycosyl hydrolase 9C2
Chromosome Location
Chr.1: 23911024 - 23915236 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105114.3 NP_176621.1 2 - 3 712 72
GenBank mRNA AF372940.1 - 2 - 3 541 72
AY143945.1 - 2 - 3 407 72

View Full Product Details