Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GH9C2
Entrez Gene ID
842747
Gene Name
glycosyl hydrolase 9C2
Gene Aliases
AT1G64390, AtGH9C2, F15H21.9, F15H21_9, glycosyl hydrolase 9C2
Chromosome Location
Chr.1: 23911024 - 23915236 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105114.3 NP_176621.1 3 - 4 917 81
GenBank mRNA AF372940.1 - 3 - 4 746 81
AY143945.1 - 3 - 4 612 81

View Full Product Details