Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UNE16
Entrez Gene ID
826998
Gene Name
Homeodomain-like superfamily protein
Gene Aliases
AT4G13640, F18A5.30, F18A5_30, unfertilized embryo sac 16
Chromosome Location
Chr.4: 7936592 - 7938705 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036549.2 NP_001031626.1 4 - 5 536 87
NM_117438.4 NP_567408.1 4 - 5 596 87
GenBank mRNA AK226443.1 - 4 - 5 454 87
AY087771.1 - 4 - 5 521 87

View Full Product Details