Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
J20
Entrez Gene ID
827017
Gene Name
DNAJ-like 20
Gene Aliases
AT4G13830, DJC26, DNA J protein C26, DNAJ-like 20, F18A5.220, F18A5_220
Chromosome Location
Chr.4: 8010967 - 8012880 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117457.5 NP_193119.1 1 - 2 1004 68
GenBank mRNA AF370236.1 - 1 - 2 542 68
AY113929.1 - 1 - 2 453 68

View Full Product Details