Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRD1
Entrez Gene ID
827058
Gene Name
putative recombination initiation defect 1
Gene Aliases
AT4G14180, AtPRD1, DL3130W, FCAALL.114, putative recombination initiation defect 1
Chromosome Location
Chr.4: 8178330 - 8184225 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340906.1 NP_001329589.1 4 - 5 1078 138

View Full Product Details