Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELIP2
Entrez Gene ID
827119
Gene Name
Chlorophyll A-B binding family protein
Gene Aliases
AT4G14690, DL3385W, EARLY LIGHT-INDUCIBLE PROTEIN 2, FCAALL.232
Chromosome Location
Chr.4: 8418242 - 8419325 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117551.3 NP_567438.1 1 - 2 261 74
GenBank mRNA AF370266.1 - 1 - 2 220 74
AY063083.1 - 1 - 2 130 74

View Full Product Details