Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPD1
Entrez Gene ID
827123
Gene Name
TIFY domain/Divergent CCT motif family protein
Gene Aliases
AT4G14713, PEAPOD 1, PLANT-SPECIFIC PUTATIVE DNA-BINDING PROTEIN 1, TIFY4A
Chromosome Location
Chr.4: 8426677 - 8429720 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117555.4 NP_567442.2 5 - 6 820 151
NM_202819.3 NP_974548.1 5 - 6 767 151
GenBank mRNA BT010167.1 - 5 - 6 625 151
KM667998.1 - 5 - 6 625 151

View Full Product Details