Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TCX2
- Entrez Gene ID
-
827132
- Gene Name
- TESMIN/TSO1-like CXC 2
- Gene Aliases
-
AT4G14770, ATTCX2, DL3425C, FCAALL.306, TESMIN/TSO1-LIKE CXC 2, TESMIN/TSO1-like CXC 2
- Chromosome Location
-
Chr.4: 8480634 - 8484976 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117562.7 |
NP_193213.5 |
1 - 2 |
215 |
100 |
View Full Product Details