Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGLYAH
Entrez Gene ID
827413
Gene Name
ureidoglycine aminohydrolase
Gene Aliases
AT4G17050, DL4555W, FCAALL.343, ureidoglycine aminohydrolase
Chromosome Location
Chr.4: 9589501 - 9592590 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117809.6 NP_193438.2 11 - 12 931 129
GenBank mRNA AK118019.1 - 11 - 12 816 129
GQ303359.1 - 11 - 12 770 129

View Full Product Details