Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAN3
Entrez Gene ID
827490
Gene Name
Homeodomain-like superfamily protein
Gene Aliases
AT4G17695, KANADI 3
Chromosome Location
Chr.4: 9847933 - 9850927 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117878.3 NP_567535.1 1 - 2 799 81
GenBank mRNA AY048690.1 - 1 - 2 578 81

View Full Product Details