Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PYR1
Entrez Gene ID
827510
Gene Name
Polyketide cyclase/dehydrase and lipid transport superfamily protein
Gene Aliases
AT4G17870, PYRABACTIN RESISTANCE 1, RCAR11, T6K21.50, T6K21_50, regulatory component of ABA receptor 11
Chromosome Location
Chr.4: 9928587 - 9929875 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117896.3 NP_193521.1 1 - 1 752 61
GenBank mRNA AY042890.1 - 1 - 1 643 61
AY081526.1 - 1 - 1 547 61

View Full Product Details