Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGD8
Entrez Gene ID
827512
Gene Name
ARF-GAP domain 8
Gene Aliases
ARF-GAP domain 8, AT4G17890, T6K21.1
Chromosome Location
Chr.4: 9936861 - 9939493 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036581.1 NP_001031658.1 3 - 4 733 75
NM_117898.5 NP_567543.1 3 - 4 896 75
GenBank mRNA AK176287.1 - 3 - 4 733 75
AK176618.1 - 3 - 4 735 75
AK221239.1 - 3 - 4 722 75
AK221371.1 - 3 - 4 722 75
AK229721.1 - 3 - 4 697 75
AK317033.1 - 3 - 4 728 75
BT000968.1 - 3 - 4 636 75

View Full Product Details