Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DEG3
- Entrez Gene ID
-
842874
- Gene Name
- DegP protease 3
- Gene Aliases
-
AT1G65630, DegP protease 3, DegP3, F1E22.1, degradation of periplasmic proteins 3
- Chromosome Location
-
Chr.1: 24406461 - 24409068 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105236.2 |
NP_564856.1 |
1 - 2 |
373 |
84 |
View Full Product Details