Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YSL7
Entrez Gene ID
842884
Gene Name
YELLOW STRIPE like 7
Gene Aliases
AT1G65730, F1E22.10, F1E22_10, YELLOW STRIPE like 7
Chromosome Location
Chr.1: 24442388 - 24446291 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105247.4 NP_176750.1 2 - 3 892 64
GenBank mRNA AY515565.1 - 2 - 3 752 64
BT002954.1 - 2 - 3 680 64
BT006077.1 - 2 - 3 641 64

View Full Product Details