Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LINC1
Entrez Gene ID
843043
Gene Name
little nuclei1
Gene Aliases
(Japanese for nucleus) 2, AT1G67230, AtLINC1, CROWDED NUCLEI 1, CRWN1, F1N21.5, KAKU2, LITTLE NUCLEI1
Chromosome Location
Chr.1: 25151211 - 25156389 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105392.5 NP_176892.1 5 - 6 1363 113

View Full Product Details