Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSM1
Entrez Gene ID
843127
Gene Name
S-adenosyl-L-methionine-dependent methyltransferases superfamily protein
Gene Aliases
AT1G67990, ATTSM1, T23K23.16, T23K23_16, TAPETUM-SPECIFIC METHYLTRANSFERASE 1
Chromosome Location
Chr.1: 25489455 - 25490867 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105469.2 NP_564917.1 3 - 4 325 76
GenBank mRNA AY088274.1 - 3 - 4 325 76
BT026412.1 - 3 - 4 286 76

View Full Product Details