Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARG1
Entrez Gene ID
843166
Gene Name
Chaperone DnaJ-domain superfamily protein
Gene Aliases
ALTERED RESPONSE TO GRAVITY 1, AT1G68370, T22E19.25, T22E19_25
Chromosome Location
Chr.1: 25631689 - 25634780 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105508.4 NP_177004.1 5 - 6 809 154
GenBank mRNA AF089810.1 - 5 - 6 717 154
BT025556.1 - 5 - 6 556 154

View Full Product Details