Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPC4
Entrez Gene ID
843206
Gene Name
phosphoenolpyruvate carboxylase 4
Gene Aliases
AT1G68750, ATPPC4, F14K14.14, F14K14_14, phosphoenolpyruvate carboxylase 4
Chromosome Location
Chr.1: 25822716 - 25828147 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105548.5 NP_177043.2 14 - 15 1651 101
GenBank mRNA AJ532903.1 - 14 - 15 1608 101

View Full Product Details