Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MES15
Entrez Gene ID
843255
Gene Name
methyl esterase 15
Gene Aliases
ARABIDOPSIS THALIANA METHYL ESTERASE 15, AT1G69240, ATMES15, F4N2.19, F4N2_19, RHS9, ROOT HAIR SPECIFIC 9, methyl esterase 15
Chromosome Location
Chr.1: 26027882 - 26030312 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105591.3 NP_177084.2 1 - 2 623 99

View Full Product Details