Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGL94
Entrez Gene ID
843289
Gene Name
AGAMOUS-like 94
Gene Aliases
AGAMOUS-like 94, AT1G69540, F10D13.25, F10D13_25
Chromosome Location
Chr.1: 26145001 - 26147751 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334413.1 NP_001322116.1 4 - 5 682 137
NM_105623.5 NP_177113.3 4 - 5 682 137
GenBank mRNA AB115013.1 - 3 - 4 259 137

View Full Product Details