Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PP2A
Entrez Gene ID
843333
Gene Name
serine/threonine protein phosphatase 2A
Gene Aliases
AT1G69960, F20P5.30, F20P5_30, TYPE 2A SERINE/THREONINE PROTEIN PHOSPHATASE, serine/threonine protein phosphatase 2A
Chromosome Location
Chr.1: 26348721 - 26350697 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105665.3 NP_177154.1 2 - 3 552 144
GenBank mRNA AK227054.1 - 2 - 3 522 144
BT025871.1 - 2 - 3 366 144
U39568.2 - 2 - 3 490 144

View Full Product Details