Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NdhL
- Entrez Gene ID
-
843413
- Gene Name
- inorganic carbon transport protein-like protein
- Gene Aliases
-
AT1G70760, CHLORORESPIRATORY REDUCTION 23, CRR23, F5A18.6, F5A18_6, NADH dehydrogenase-like complex L
- Chromosome Location
-
Chr.1: 26687202 - 26688384 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105744.4 |
NP_177233.1 |
1 - 2 |
186 |
92 |
GenBank mRNA |
BT010638.1 |
- |
1 - 2 |
121 |
92 |
View Full Product Details