The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198441.1 | NP_001185370.1 | 17 - 18 | 2300 | 107 |
NM_001334490.1 | NP_001323059.1 | 17 - 18 | 2380 | 107 | |
NM_105791.5 | NP_177278.3 | 17 - 18 | 2300 | 107 | |
GenBank mRNA | AK230327.1 | - | 17 - 18 | 2300 | 107 |