Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOL1
Entrez Gene ID
843499
Gene Name
carboxypeptidase D
Gene Aliases
AT1G71696, F14O23.7, F14O23_7, SUPPRESSOR OF LLP1 1
Chromosome Location
Chr.1: 26966766 - 26970603 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105827.3 NP_177314.2 11 - 12 1391 98
NM_202397.2 NP_974126.1 12 - 13 1249 98
GenBank mRNA AJ555410.1 - 12 - 13 1128 98

View Full Product Details