Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P40
Entrez Gene ID
843569
Gene Name
40s ribosomal protein SA
Gene Aliases
40s ribosomal protein SA, AP40, AT1G72370, RP40, RPSAA, T10D10.16, T10D10_16
Chromosome Location
Chr.1: 27242845 - 27244931 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105896.4 NP_177381.1 4 - 5 919 57
GenBank mRNA AY054211.1 - 4 - 5 895 57
AY058885.1 - 4 - 5 915 57
AY065096.1 - 4 - 5 912 57
AY079041.1 - 4 - 5 830 57
AY087976.1 - 4 - 5 912 57
AY114561.1 - 4 - 5 830 57
AY136324.1 - 4 - 5 892 57
BT000421.1 - 4 - 5 830 57
U01955.1 - 4 - 5 884 57

View Full Product Details