Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDG11
Entrez Gene ID
843671
Gene Name
homeodomain GLABROUS 11
Gene Aliases
AT1G73360, ATHDG11, AtEDT1, EDT1, ENHANCED DROUGHT TOLERANCE 1, HOMEODOMAIN GLABROUS 11, T9L24.43, T9L24_43, homeodomain GLABROUS 11
Chromosome Location
Chr.1: 27578421 - 27582685 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105996.4 NP_177479.1 1 - 2 1005 77
GenBank mRNA BT003979.1 - 1 - 2 746 77
BT004915.1 - 1 - 2 140 77

View Full Product Details