Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AAO2
- Entrez Gene ID
-
823457
- Gene Name
- aldehyde oxidase 2
- Gene Aliases
-
ALDEHYDE OXIDASE 2, AO3, AOgamma, AT3G43600, Aldehyde oxidase gamma, Arabidopsis thaliana aldehyde oxidase 2, Arabidopsis thaliana aldehyde oxidase 3, AtAO3, aldehyde oxidase 2, aldehyde oxidase 3, atAO-2
- Chromosome Location
-
Chr.3: 15511832 - 15517545 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_114228.3 |
NP_189946.1 |
3 - 4 |
2341 |
81 |
GenBank mRNA |
AB005805.1 |
- |
3 - 4 |
2302 |
81 |
View Full Product Details