Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GH9A4
Entrez Gene ID
823499
Gene Name
glycosyl hydrolase 9A4
Gene Aliases
AT3G43860, AtGH9A4, glycosyl hydrolase 9A4
Chromosome Location
Chr.3: 15706954 - 15709989 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339113.1 NP_001327805.1 1 - 2 457 68
NM_114254.3 NP_189972.2 1 - 2 457 68
GenBank mRNA AY072099.1 - 1 - 2 417 68

View Full Product Details