Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP71
Entrez Gene ID
823585
Gene Name
cyclophilin71
Gene Aliases
AT3G44600, AtCYP71, cyclophilin 71, cyclophilin71
Chromosome Location
Chr.3: 16164935 - 16169304 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114328.4 NP_190046.2 5 - 6 827 115
GenBank mRNA AY062653.1 - 5 - 6 785 115
BT008382.1 - 5 - 6 724 115

View Full Product Details